Skip to Content
Merck
All Photos(1)

Key Documents

664111

Sigma-Aldrich

2-Fluoro-3-pyridinecarboxaldehyde

97%

Synonym(s):

2-Fluoro-3-formylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4FNO
CAS Number:
Molecular Weight:
125.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.520

bp

50 °C/3 mmHg

density

1.250 g/mL at 25 °C

SMILES string

[H]C(=O)c1cccnc1F

InChI

1S/C6H4FNO/c7-6-5(4-9)2-1-3-8-6/h1-4H

InChI key

OFBVGDCXXGXDKU-UHFFFAOYSA-N

Application

Attachment of isoxazolinones by displacement of fluoride provide naphthyridines on hydrogenolysis and cyclization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 59, 9887-9887 (2003)
Olivia Morris et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 116, 120-127 (2016-08-16)
2-[(18)F]-Fluoro-3-pyridinecarboxaldehyde ([(18)F]FPCA) is a novel, water-soluble prosthetic group. It's radiochemistry has been developed and fully-automated for application in chemoselective radiolabelling of amino(oxy)-derivatised RI-OR2-TAT peptide, (Aoa-k)-RI-OR2-TAT, using a GE TRACERlab FX-FN. RI-OR2-TAT is a brain-penetrant, retro-inverso peptide that binds to amyloid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service