Skip to Content
Merck
All Photos(1)

Key Documents

639893

Sigma-Aldrich

Glycidaldehyde diethyl acetal

90%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€133.00

€133.00


Estimated to ship on21 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€133.00

About This Item

Empirical Formula (Hill Notation):
C7H14O3
CAS Number:
Molecular Weight:
146.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€133.00


Estimated to ship on21 April 2025


Request a Bulk Order

Quality Level

Assay

90%

refractive index

n20/D 1.414 (lit.)

density

0.943 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CCOC(OCC)C1CO1

InChI

1S/C7H14O3/c1-3-8-7(9-4-2)6-5-10-6/h6-7H,3-5H2,1-2H3

InChI key

QIQQWSQARNQPLP-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Fu et al.
Free radical biology & medicine, 73, 12-20 (2014-05-13)
Previous studies showed that 7-(1',2'-dihydroxyheptyl)-substituted etheno DNA adducts are products of reactions with the epoxide of (E)-4-hydroxy-2-nonenal, an oxidation product of ω-6 polyunsaturated fatty acids (PUFAs). In this work, we report the detection of 7-(1',2'-dihydroxyheptyl)-1,N(6)-ethenodeoxyadenosine (DHHedA) in rodent and human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service