Skip to Content
Merck
All Photos(1)

Key Documents

634417

Sigma-Aldrich

Yttrium iron oxide

Yttrium iron oxide

nanopowder, <100 nm particle size (BET), 99.9% trace metals basis

Synonym(s):

Iron yttrium oxide, YIG, Yttrium ferrite garnet, Yttrium iron garnet

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y3Fe5O12
CAS Number:
Molecular Weight:
737.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

nanopowder

surface area

20-40 m2/g

particle size

<100 nm (BET)

density

5.11 g/mL at 25 °C (lit.)

SMILES string

O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Y]=O.O=[Y]O[Y]=O

InChI

1S/5Fe.12O.3Y

InChI key

UVXIKKWNYGPENJ-UHFFFAOYSA-N

Application

Magnetic garnet with interesting optical and magnetic properties.[1]

Legal Information

Product of Engi-Mat Co.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baolin Wu et al.
Statistics and its interface, 10(3), 379-386 (2017-02-22)
More and more large cohort studies have conducted or are conducting genome-wide association studies (GWAS) to reveal the genetic components of many complex human diseases. These large cohort studies often collected a broad array of correlated phenotypes that reflect common
Rastogi, A.C.; Moorthy, V.N.
Materials Science and Engineering, B, 95, 131-131 (2002)
Yttrium oxide nanoparticles prepared by heat treatment of cathodically grown yttrium hydroxide
Aghazadeh M, et al.
ISRN Ceramics (2011)
Anne Rifkin-Graboi et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(34), 16787-16792 (2019-08-07)
Attachment disorganization is a risk factor for difficulties in attention, social relationships, and mental health. Conceptually, attachment disorganization may indicate a breakdown in fear regulation resulting from repeated exposure to frightening maternal care. In addition, past research has examined the
Tim Schmitz et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 18(9), 1125-1132 (2016-12-30)
Pulsed laser ablation of pressed yttrium iron garnet powders in water is studied and compared to the ablation of a single-crystal target. We find that target porosity is a crucial factor, which has far-reaching implications on nanoparticle productivity. Although nanoparticle

Articles

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

Magnetic materials find diverse applications from data storage to renewable energy.

Questions

  1. Can you please email me the XRD that was collected for Y3Fe5O12, product number 634417, batch number MKCN0637.

    1 answer
    1. Unfortunately, the XRD is not available for this product.

      Helpful?

Reviews

Active Filters

  1. Canada
    • Review 1
    • Vote 1
    1 out of 5 stars.

    This powder has a lot of orthoferrite YFO3 phase instead of Y3Fe5O12 main phase!

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service