Skip to Content
Merck
All Photos(1)

Key Documents

50642

Sigma-Aldrich

Glycolic acid nitrile solution

~55% in H2O

Synonym(s):

Formaldehyde cyanohydrin, Hydroxyacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CN
CAS Number:
Molecular Weight:
57.05
Beilstein:
605328
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

contains

~0.5% phosphoric acid as stabilizer

concentration

~55% in H2O

refractive index

n20/D 1.376

SMILES string

OCC#N

InChI

1S/C2H3NO/c3-1-2-4/h4H,2H2

InChI key

LTYRAPJYLUPLCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Arrhenius et al.
The Journal of organic chemistry, 62(16), 5522-5525 (1997-08-08)
A study of glycolonitrile polymerization has led to the isolation and characterization of two 2,5-dihydro-4-aminooxazoles, 4 and 5. Previous reports have misassigned these structures as s-triazines or pyrimidines. X-ray diffraction analysis of crystals of 4 and an acetylated oxazole derivative
Shijun Wu et al.
Biotechnology and bioengineering, 99(3), 717-720 (2007-09-06)
A key step in a chemoenzymatic process for the production of high-purity glycolic acid (GLA) is the enzymatic conversion of glycolonitrile (GLN) to ammonium glycolate using a nitrilase derived from Acidovorax facilis 72W. Protein engineering and over-expression of this nitrilase
Evidence for the microsomal metabolism of glycolonitrile.
J J Freeman et al.
Biochemical pharmacology, 36(1), 184-187 (1987-01-01)
Acceleration of HCN oligomerization by formaldehyde and related compounds: implications for prebiotic syntheses.
A W Schwartz et al.
Journal of molecular evolution, 18(5), 351-353 (1982-01-01)
P Eyer et al.
Archives of toxicology, 59(4), 266-271 (1986-12-01)
HI 6 has been shown to be efficacious in soman intoxication of laboratory animals by reactivation of acetylcholinesterase. To assess possible risks involved in the administration of HI 6 its degradation products were analyzed at pH 2.0, 4.0, 7.4, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service