Skip to Content
Merck
All Photos(1)

Key Documents

496642

Sigma-Aldrich

3-Cyanophenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4NCS
CAS Number:
Molecular Weight:
160.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

64-66 °C (lit.)

functional group

isothiocyanate
nitrile

storage temp.

2-8°C

SMILES string

S=C=Nc1cccc(c1)C#N

InChI

1S/C8H4N2S/c9-5-7-2-1-3-8(4-7)10-6-11/h1-4H

InChI key

UXCYPHCFQAFVFH-UHFFFAOYSA-N

General description

3-Cyanophenyl isothiocyanate, also known as 3-isothiocyanatobenzonitrile, is an organic building block containing an isocyanate group Its freezing point, density and refractive index have been reported.

Application

3-Cyanophenyl isothiocyanate may be used in the synthesis of ((3-cyanophenyl)amino)((2-phenylethyl)amino)methane- 1-thione and N-((3-cyanophenyl)carbamothioyl)benzimidamide.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-catalyzed synthesis of 3-substituted-5-amino-1,2,4-thiadiazoles via intramolecular N-S bond formation.
Kim HY, et al.
Tetrahedron, 70(45), 8737-8743 (2014)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 178-178 (2015)
Preparation of aryl isothiocyanates via protected phenylthiocarbamates and application to the synthesis of caffeic acid (4-isothiocyanato)phenyl ester.
X Zhang et al.
The Journal of organic chemistry, 65(19), 6237-6240 (2000-09-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service