Skip to Content
Merck
All Photos(2)

Key Documents

481076

Sigma-Aldrich

Diphenylzinc

92%

Synonym(s):

Diphenylzinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2Zn
CAS Number:
Molecular Weight:
219.60
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

92%

form

powder or crystals

impurities

<7.5% benzene

bp

280-285 °C (lit.)

mp

102-106 °C (lit.)

SMILES string

[Zn](c1ccccc1)c2ccccc2

InChI

1S/2C6H5.Zn/c2*1-2-4-6-5-3-1;/h2*1-5H;

InChI key

MKRVHLWAVKJBFN-UHFFFAOYSA-N

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Muta. 1B - Pyr. Sol. 1 - STOT RE 2

Target Organs

Blood

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service