Skip to Content
Merck
All Photos(1)

Documents

452416

Sigma-Aldrich

Decane-d22

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)8CD3
CAS Number:
Molecular Weight:
164.42
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.407 (lit.)

bp

174 °C (lit.)

mp

−30 °C (lit.)

density

0.842 g/mL at 25 °C

mass shift

M+22

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2

InChI key

DIOQZVSQGTUSAI-DNYWURFXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F

Flash Point(C)

46 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian Paz Ramos et al.
Biochimica et biophysica acta. Biomembranes, 1862(5), 183201-183201 (2020-01-24)
Alkanes are known to promote the fluid lamellar (Lα)-to-inverted hexagonal (HII) phase transition of different phospholipids. In this work, we studied the interaction of decane and tetradecane with self-assemblies formed of 1-palmitoyl-2-oleoyl-sn-glycero-phosphoethanolamine (POPE), using sequential 2H and 31P solid-state NMR spectroscopy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service