Skip to Content
Merck
All Photos(1)

Documents

428167

Sigma-Aldrich

Chloric acid solution

35 wt. %

Synonym(s):

Chloric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO3
CAS Number:
Molecular Weight:
84.46
MDL number:
UNSPSC Code:
12000000

concentration

35 wt. %

bp

>100 °C

density

1.2 g/mL at 25 °C

SMILES string

OCl(=O)=O

General description

Chloric acid solution (Chloric acid) is a strong oxidizing agent. It can be synthesized by reacting barium chlorate with dilute sulfuric acid. Chloric acid is reported to be formed during the decompostion of aqueous solution of chlorine dioxide.

Application

Chloric acid solution (Chloric acid) may be used in the synthesis of crystals of anhydrous mercurous chlorate.
Stable precursor to chlorine dioxide. Used in electrochemical production of high-purity ammonium perchlorate.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Liq. 1 - Skin Corr. 1B

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dotson, R.L.
J. Appl. Electrochem., 23, 897-897 (1993)
Anhydrous Mercurous Chlorate, Hg2(ClO3)2.
Goebbels D and Meyer G.
Zeitschrift fur Anorganische und Allgemeine Chemie, 629(14), 2446-2447 (2003)
The Use of Chlorine Dioxide in Water Treatment.
Vincent GP, et al.
American Journal of Public Health and the nation's Health, 36(9), 1035-1037 (1946)
Eagleson M.
Concise Encyclopedia Chemistry, 212-213 (1994)
Agnieszka Robaszkiewicz
Postepy biochemii, 56(2), 201-208 (2010-09-29)
Reactive oxygen and nitrogen species are attributed to initiation and propagation of many diseases. The demonstration of elevated activity of myeloperoxidase and the level of 3-chlorotyrosine in atherosclerosis, kidney diseases and chronic inflammations brought about the interest in the biological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service