Skip to Content
Merck
All Photos(1)

Key Documents

399507

Sigma-Aldrich

Niobium diboride

−325 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NbB2
CAS Number:
Molecular Weight:
114.53
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

particle size

−325 mesh

density

6.97 g/mL at 25 °C (lit.)

SMILES string

B#[Nb]#B

InChI

1S/2B.Nb

InChI key

WCJIBJJSTRKWNY-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service