Skip to Content
Merck
All Photos(1)

Key Documents

39740

Sigma-Aldrich

2,2-Dimethylbutane

≥99.0% (GC)

Synonym(s):

Neohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C(CH3)3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730736
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.97 (vs air)

vapor pressure

5.35 psi ( 20 °C)

Assay

≥99.0% (GC)

autoignition temp.

797 °F

expl. lim.

~7.7 %

refractive index

n20/D 1.369 (lit.)

bp

50 °C (lit.)

mp

−100 °C (lit.)

density

0.649 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCC(C)(C)C

InChI

1S/C6H14/c1-5-6(2,3)4/h5H2,1-4H3

InChI key

HNRMPXKDFBEGFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2-Dimethylbutane (Neohexane) is an acyclic alkane. It is an isomeric form of hexane. Its Research Octane Number (RON) has been reported to be 91.8. Oxidation of 2,2-dimethylbutane at low temperatures has been studied in a jet-stirred reactor (JSR). Pt-Re catalyzed reactions of 2,2-dimethylbutane (neohexane) has been studied.

Application

2,2-Dimethylbutane may be employed as probe to investigate the nature of possible active sites in supported metal catalysts.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhandong Wang et al.
The journal of physical chemistry. A, 118(30), 5573-5594 (2014-07-10)
The low-temperature oxidation of the five hexane isomers (n-hexane, 2-methyl-pentane, 3-methyl-pentane, 2,2-dimethylbutane, and 2,3-dimethylbutane) was studied in a jet-stirred reactor (JSR) at atmospheric pressure under stoichiometric conditions between 550 and 1000 K. The evolution of reactant and product mole fraction
Hydrogenolysis of 2,2-dimethylbutane and n-hexane over supported ruthenium, nickel, cobalt, and iron.
Machiels CJ and Anderson RB.
J. Catal., 58(2), 268-275 (1979)
The use of 2,2-dimethylbutane (neohexane) as a probe molecule of metal catalysts.
Burch R and Paal Z.
Applied Catalysis A: General, 114(1), 9-33 (1994)
The effect of alloying Pt with Re on the intermediates in hydrocarbon reactions: Reactions of 2, 2-dimethylbutane.
Botman MJP, et al.
J. Catal., 116(2), 467-479 (1989)
Yohan Lee et al.
Environmental science & technology, 49(14), 8899-8906 (2015-06-25)
The CH4/CO2 replacement that occurs in sH hydrates is investigated, with a primary focus on the enhanced CH4 recovery induced via structural transformation with a CO2 injection. In this study, neohexane (NH) is used as a liquid hydrocarbon guest in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service