Skip to Content
Merck
All Photos(1)

Key Documents

268445

Sigma-Aldrich

Isethionic acid ammonium salt

99%

Synonym(s):

2-Hydroxyethanesulfonic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2SO3NH4
CAS Number:
Molecular Weight:
143.16
Beilstein:
3722656
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

138-141 °C (lit.)

functional group

hydroxyl
sulfonic acid

SMILES string

N.OCCS(O)(=O)=O

InChI

1S/C2H6O4S.H3N/c3-1-2-7(4,5)6;/h3H,1-2H2,(H,4,5,6);1H3

InChI key

LLOHIFXFHGMBNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isethionic acid ammonium salt may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Zhang et al.
Biosensors & bioelectronics, 24(4), 690-695 (2008-08-19)
A composite film of polyaniline (PAN) nano-networks/p-aminobenzene sulfonic acid (ABSA) modified glassy carbon electrode (GCE) has been fabricated via an electrochemical oxidation procedure and applied to the electro-catalytic oxidation of uric acid (UA) and ascorbic acid (AA). The ABSA monolayer
M Syringas et al.
Molecular pharmacology, 58(6), 1404-1411 (2000-11-28)
Catecholamine transporters constitute the biological targets for several important drugs, including antidepressants, cocaine, and related compounds. Some information exists about discrete domains of these transporters that are involved in substrate translocation and uptake blockade, but delineation of domains mediating the
Sonja Weinitschke et al.
Applied and environmental microbiology, 76(2), 618-621 (2009-11-26)
Ubiquitous isethionate (2-hydroxyethanesulfonate) is dissimilated by diverse bacteria. Growth of Cupriavidus necator H16 with isethionate was observed, as was inducible membrane-bound isethionate dehydrogenase (IseJ) and inducible transcription of the genes predicted to encode IseJ and a transporter (IseU). Biodiversity in
Saswata Ghosh et al.
Journal of cosmetic science, 58(3), 229-244 (2007-06-29)
Sodium cocoyl isethionate (SCI) is an important surfactant ingredient in mild, syndet (synthetic detergent) cleansing bars. In vitro and in vivo studies have demonstrated that SCI is mild and less damaging to the skin barrier than soaps and surfactants such
C Nabel et al.
Pflugers Archiv : European journal of physiology, 438(5), 694-699 (1999-11-11)
The aim of this study was to assess the relevance of chloride channels to the inhibitory effect of angiotensin II (ANGII) on renin secretion. We thus examined the effects of the chloride channels blockers IAA-94 and niflumic acid, the Na-K-Cl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service