Skip to Content
Merck
All Photos(4)

Key Documents

233390

Sigma-Aldrich

Lindane

97%

Synonym(s):

γ-BHC, 1α,2α,3β,4α,5α,6β-Hexachlorocyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83
Beilstein:
1907337
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

113-115 °C (lit.)

solubility

chloroform: soluble 50 mg/mL
water: soluble 8.35 g/L at 25 °C

SMILES string

Cl[C@H]1[C@H](Cl)[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@@H]1Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5+,6+

InChI key

JLYXXMFPNIAWKQ-GNIYUCBRSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

  • Lindane is an organochlorine pesticide and has been investigated in the water and sediments to evaluate their current pollution levels and potential risks.
  • Influence of temperature, moisture and pH on degradation and degradation kinetics of lindane during vegetable waste composting using full-scale continuous rotary drum composter (FSCRDC) has been reported.
  • Photo-oxidation of lindane in aqueous solution with titanium dioxide as the photocatalyst has been investigated.
  • Degradation of lindane by nanoscale zero-valent iron has been investigated.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degradation of lindane by zero-valent iron nanoparticles.
Elliott DW, et al.
Journal of Environmental Engineering (New York, N.Y.), 135(5), 317-324 (2009)
M Muñoz-Morales et al.
Journal of environmental management, 262, 110364-110364 (2020-04-07)
In this work, a three-step process (adsorption-desorption-electrolysis) is evaluated as an interesting approach for the removal of organochlorinated compounds (clopyralid, lindane and perchloroethylene) with different physical properties (solubility and vapor pressure) from low concentrated wastewater. First steps are based on
Muntjeer Ali et al.
Chemosphere, 102, 68-75 (2014-01-29)
Study focused on effects of temperature, moisture and pH on degradation and degradation kinetics of aldrin, endosulfan (α), endosulfan (β) and lindane during vegetable waste composting using full-scale continuous rotary drum composter (FSCRDC). Extraction, concentration and quantification of pesticides were
Ying Hu et al.
Environmental science and pollution research international, 21(15), 9376-9384 (2014-04-18)
Seventeen organochlorine pesticides (OCPs) were investigated in the water and sediments from a waterbird-inhabited lake (Yangchaihu Lake) to evaluate their current pollution levels and potential risks. The concentrations of total OCPs in water and sediments were 10.12-59.75 ng/l and 4.25-27.35
Photo-oxidation of organic compound in the presence of titanium dioxide: determination of the efficiency.
Sabin F, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 63(1), 99-106 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service