Skip to Content
Merck
All Photos(1)

Documents

217344

Sigma-Aldrich

Trichloroacetamide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCONH2
CAS Number:
Molecular Weight:
162.40
Beilstein:
1754028
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

bp

238-240 °C (lit.)

mp

139-141 °C (lit.)

SMILES string

NC(=O)C(Cl)(Cl)Cl

InChI

1S/C2H2Cl3NO/c3-2(4,5)1(6)7/h(H2,6,7)

InChI key

UPQQXPKAYZYUKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloroacetamide is the major degradation product of trichloroacetonitrile.

Application

Trichloroacetamide was used in microarray-based transcriptomics and one-dimensional proton nuclear magnetic resonance based metabonomics to investigate the health effects of nitrogenous disinfection byproducts of trichloroacetamide in mice.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J Meloncelli et al.
Carbohydrate research, 346(12), 1406-1426 (2011-05-03)
The ABO histo-blood group antigens are best known for their important roles in solid organ and bone marrow transplantation as well as transfusion medicine. Here we report the synthesis of the ABO type III and IV antigens with a 7-octen-1-yl
Jeffrey S Arnold et al.
Chemical communications (Cambridge, England), 48(94), 11531-11533 (2012-10-24)
We report the chiral diene ligated rhodium-catalyzed dynamic kinetic asymmetric transformation (DYKAT) of racemic secondary allylic trichloroacetimidates with a variety of N-methyl anilines, providing allylic N-methyl arylamines in high yields, regioselectivity, and enantiomeric excess. The rhodium-catalyzed DYKAT method addresses limitations
Jeffrey S Arnold et al.
Organic letters, 12(20), 4580-4583 (2010-09-17)
The use of unactivated aromatic amines in the rhodium-catalyzed regioselective amination of secondary allylic trichloroacetimidates is explored. The desired N-arylamines are obtained in high yields and regioselectivity, favoring the branched amination products. The presence of the trichloroacetimidate leaving group was
Yan Zhang et al.
Environmental science & technology, 47(6), 2918-2924 (2013-02-15)
Microarray-based transcriptomics and one-dimensional proton nuclear magnetic resonance ((1)H NMR) based metabonomics approaches were employed to investigate the health effects of nitrogenous disinfection byproducts (N-DBPs) of trichloroacetamide (TCAcAm) on mice. Mice were exposed to TCAcAm at concentrations of 50, 500
Nihan Celebi-Olçüm et al.
The Journal of organic chemistry, 74(18), 6944-6952 (2009-08-20)
Density functional theory calculations were used to investigate the [3,3]- and [1,3]-shifts of O-allylic trichloroacetimidates in the presence of cinchona alkaloids. Thermal [1,3]- and [3,3]-rearrangements proceed through concerted pseudopericyclic transition states to give the corresponding rearranged products. [1,3]-Rearrangement is catalyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service