Skip to Content
Merck
All Photos(1)

Key Documents

187135

Sigma-Aldrich

Isonicotinic acid N-oxide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

270-271 °C (lit.)

SMILES string

OC(=O)c1cc[n+]([O-])cc1

InChI

1S/C6H5NO3/c8-6(9)5-1-3-7(10)4-2-5/h1-4H,(H,8,9)

InChI key

QCWTWMJMLSKQCJ-UHFFFAOYSA-N

General description

Isonicotinic acid N-oxide interacts with 3d metal(II) perchlorates in ethanol-triethyl orthoformate leading to partial substitution of perchlorate with isonicotinate-N-oxide anionic groups. It reacts with AgClO4 or AgBF4 to yield distinct crystalline products. It forms polymeric lanthanide(III) complexes.

Application

Isonicotinic acid N-oxide was used in the synthesis of three-dimensional coordination polymers based on inorganic lanthanide(II) sulfate skeletons.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicotinic and isonicotinic acid n-oxide interactions with 3d metal perchlorates [1].
Gelfand LS, et al.
J. Inorg. Nucl. Chem., 42(3), 377-385 (1980)
Structural characterization and luminescence studies of new lanthanide (III) complexes with nicotinic and isonicotinic acid N-oxides.
Mao J-G, et al.
Polyhedron, 17(23), 3999-4009 (1998)
Nasrin Shokouhfar et al.
Dalton transactions (Cambridge, England : 2003), 47(41), 14549-14555 (2018-09-27)
Considering the widespread production of Cr(vi) from various industrial processes and its damaging toxicity as a carcinogenic agent, it is imperative to investigate stable and efficient adsorbents with a rapid performance towards Cr(vi) adsorption. Zirconium-containing metal-organic frameworks (Zr-MOFs) have been
Unusual anion effect on the direction of three-dimensional (3-D) channel-like silver (I) coordination frameworks with isonicotinic acid N-oxide.
Du M, et al.
CrystEngComm, 11(8), 1536-1540 (2009)
Zheng He et al.
Inorganic chemistry, 44(4), 862-874 (2005-04-30)
Fourteen three-dimensional coordination polymers of general formula [Ln(lNO)(H2O)(SO4)]n, where Ln = La, 1.La; Ce, 2.Ce; Pr, 3.Pr; Nd, 4.Nd; Sm, 5.Sm; Eu, 6.Eu; Gd, 7.Gd; Tb, 8.Tb; Dy, 9.Dy; Ho, 10.Ho; Er. 11.Er; Tm, 12.Tm; Yb, 13.Yb; and Lu, 14.Lu;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service