Skip to Content
Merck
All Photos(1)

Key Documents

AC7300012789

Advanced Chemical Intermediates Ltd.

N-Hydroxy-4-methoxybenzenecarboximidoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
185.61
MDL number:

Iupac Name

N-hydroxy-4-methoxybenzenecarboximidoyl chloride

Inchi Code

1S/C8H8ClNO2/c1-12-7-4-2-6(3-5-7)8(9)10-11/h2-5,8H,1H3

InChI key

LXKQZUDNNOFDPZ-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Physical Form

off white solid

Product Link

https://www.acints.com/products/SB-03D-050

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.acints.com/products/SB-03D-050


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service