Skip to Content
Merck
All Photos(1)

Key Documents

R7023

Sigma-Aldrich

Pst I from Providencia stuartii

Restriction Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

grade

Molecular Biology
for molecular biology

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Specificity

Recognition sequence: 5′-CTGCA/G-3′
Cutting results: a 2-10-fold Pst I overdigestion of 1 μg λ DNA substrate results in 100% cutting.
Heat inactivation: Inactivated at 80 °C for 20 minutes.

Application

PstI is a restiction enzyme used in molecular biology applications to cleave DNA at the recognition sequence 5′-CTGCA/G-3′, generating DNA fragments with 3′- cohesive termini.

Other Notes

Supplied with 10x Restriction Enzyme Buffer SH (B3657).
Comment: Pst I requires optimal reaction conditions in order to avoid star activity.

Physical form

Solution in 10 mM Tris-HCl, pH 7.0, 1 mM EDTA, 250 mM NaCl, 1 mM 2-mercaptoethanol, 50% glycerol (v/v), 0.01% polydocanol (v/v) at 4 °C

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mapping and sequence determination of the single site in phiX174am3 replicative form DNA cleaved by restriction endonuclease Pst I.
N L Brown et al.
FEBS letters, 65(3), 284-287 (1976-06-15)
E Müller et al.
Pflugers Archiv : European journal of physiology, 435(5), 705-712 (1998-04-16)
The influence of diuresis and antidiuresis on the expression of heat shock proteins (HSP) 25, 60, 72 and 73 in the renal cortex and outer and inner medulla of Wistar rats was analysed. Medullary osmolality was reduced by long-term diuresis
Thomas Hiesberger et al.
The Journal of clinical investigation, 113(6), 814-825 (2004-04-07)
Hepatocyte nuclear factor-1beta (HNF-1beta) is a Pit-1, Oct-1/2, UNC-86 (POU)/homeodomain-containing transcription factor that regulates tissue-specific gene expression in the liver, kidney, and other organs. Humans with autosomal dominant mutations of HNF-1beta develop maturity-onset diabetes of the young type 5 (MODY5)
D I Smith et al.
Nucleic acids research, 3(2), 343-353 (1976-02-01)
We describe the isolation of a new class 2 restriction endonuclease from Providencia stuartii 164. Using the procedure of osmotic shock treatment, we have partially purified this enzyme (Pst 1) and have begun preliminary work to characterize its specificity and
T B Britschgi et al.
Applied and environmental microbiology, 57(6), 1707-1713 (1991-06-01)
The identification of the prokaryotic species which constitute marine bacterioplankton communities has been a long-standing problem in marine microbiology. To address this question, we used the polymerase chain reaction to construct and analyze a library of 51 small-subunit (16S) rRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service