Skip to Content
Merck
All Photos(1)

Key Documents

N2128

Sigma-Aldrich

4-Nitroaniline

crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NH2
CAS Number:
Molecular Weight:
138.12
Beilstein:
508690
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.5%

form

crystalline

color

yellow

bp

260 °C/100 mmHg (lit.)

mp

146-149 °C (lit.)

ε (extinction coefficient)

8.8 at 410 nm at 1 mM (pH 5.0-10.5)

SMILES string

Nc1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H6N2O2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H,7H2

InChI key

TYMLOMAKGOJONV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Product of chromogenic reactions.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

415.4 °F - closed cup

Flash Point(C)

213.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cuiping Han et al.
The Analyst, 135(3), 583-588 (2010-02-23)
The purpose of this paper is to develop a sensitive and low-cost colorimetric method for detection of melamine using simple circuitry. The stable p-nitroaniline-modified silver nanoparticles (Ag NPs) were synthesized by a zero-length covalent coupling chemistry, which showed sensitivity and
Azeem Khalid et al.
Water research, 43(4), 1110-1116 (2008-12-31)
Environmentally toxic aromatic amines including nitroanilines are commonly generated in dye contaminated wastewater in which azo dyes undergo degradation under anaerobic conditions. The aim of this study was to develop a process for biological treatment of 4-nitroaniline. Three bacteria identified
Shoichi Yamaguchi et al.
The Journal of chemical physics, 129(10), 101102-101102 (2008-12-03)
Heterodyne-detected electronic sum frequency generation (HD-ESFG) spectroscopy is newly developed to obtain complex electronic chi(2) spectra of interfaces for a simultaneous detection bandwidth broader than 100 nm. HD-ESFG provides linear chi(2) spectra that have unambiguous information on the "up" versus
Krzysztof Iwaszczuk et al.
Optics express, 17(24), 21969-21976 (2009-12-10)
We present a new method for data acquisition in time-resolved terahertz spectroscopy experiments. Our approach is based on simultaneous collection of reference and differential THz scans. Both the optical THz generation beam and the pump beam are modulated at two
Kumi Y Inoue et al.
Innate immunity, 18(2), 343-349 (2011-08-17)
Here, we report the development of an electrochemical detection method for endotoxin based on the Limulus amebocyte lysate (LAL) assay. A mixture of LAL reagent and endotoxin sample solution was incubated for 1 h. The endotoxin activated a cascade reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service