Skip to Content
Merck
All Photos(1)

Key Documents

F3379

Sigma-Aldrich

Fibrinopeptide B human

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H93N19O25
CAS Number:
Molecular Weight:
1552.56
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

human

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C(NC(=O)CNC(=O)C1CCC(=O)N1)C(=O)NC(CC(N)=O)C(=O)NC(CC(O)=O)C(=O)NC(CC(N)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NCC(=O)NC(Cc2ccccc2)C(=O)NC(Cc3ccccc3)C(=O)NC(CO)C(=O)NC(C)C(=O)NC(CCCNC(N)=N)C(O)=O

InChI

1S/C66H93N19O25/c1-31(2)53(85-49(91)29-73-55(99)35-16-19-47(89)75-35)64(108)83-42(26-46(68)88)61(105)82-43(27-52(96)97)62(106)81-41(25-45(67)87)60(104)78-37(18-21-51(94)95)57(101)77-36(17-20-50(92)93)56(100)72-28-48(90)76-39(23-33-11-6-4-7-12-33)58(102)80-40(24-34-13-8-5-9-14-34)59(103)84-44(30-86)63(107)74-32(3)54(98)79-38(65(109)110)15-10-22-71-66(69)70/h4-9,11-14,31-32,35-44,53,86H,10,15-30H2,1-3H3,(H2,67,87)(H2,68,88)(H,72,100)(H,73,99)(H,74,107)(H,75,89)(H,76,90)(H,77,101)(H,78,104)(H,79,98)(H,80,102)(H,81,106)(H,82,105)(H,83,108)(H,84,103)(H,85,91)(H,92,93)(H,94,95)(H,96,97)(H,109,110)(H4,69,70,71)

InChI key

MYRIFIVQGRMHRF-UHFFFAOYSA-N

Gene Information

human ... FGB(2244)

Amino Acid Sequence

Glp-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg

General description

Fibrinopeptide B (FPB) is produced during the cleavage of fibrinogen, by thrombin, to fibrin monomer. It is cleaved off from the N-terminal of the fibrinogen β chain. This peptide is composed of 14 amino acids.

Biochem/physiol Actions

Cleavage of fibrinopeptide B from fibrinogen Bβ chains exposes E domain polymerization sites, E(B), that interact with platelets, fibroblasts and endothelial cells.
Fibrin formation is an essential part in wound healing and inflammation. Fibrinopeptide B (FPB) can also cause chemotactic migration of neutrophils, without the simultaneous release of lysosome enzymes. It is produced during the coagulation of fibrinogen, which is essential for physiological homeostasis. It is involved in various disorders such as thrombosis and disseminated intravascular coagulation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Vindigni et al.
Biochemistry, 35(14), 4417-4426 (1996-04-09)
The release of fibrinopeptides A and B by the slow and fast forms of thrombin was studied over the temperature range from 5 to 45 degrees C and the salt concentration range from 100 to 800 mM. The sequential mechanism
Heinz Nika et al.
Journal of biomolecular techniques : JBT, 24(1), 17-31 (2013-04-02)
A sample preparation method for protein C-terminal peptide isolation has been developed. In this strategy, protein carboxylate glycinamidation was preceded by carboxyamidomethylation and optional α- and ϵ-amine acetylation in a one-pot reaction, followed by tryptic digestion of the modified protein.
Bi-Ying Wei et al.
Journal of chromatography. A, 1217(44), 6927-6931 (2010-09-21)
Liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS) with selective reaction monitoring (SRM) is a selective and sensitive method for quantitation of peptides. SRM is achieved via MS/MS utilizing collision-induced dissociation (CID) while monitoring unique precursor-product ion transitions. Low-energy CID
Ernst Pittenauer et al.
Journal of proteomics, 74(7), 975-981 (2010-10-26)
The desorption/ionization behavior of individual peptides, an equimolare peptide mixture and a tryptic digest was investigated by AP-MALDI-IT-MS using four different target materials (gold-covered stainless steel (SS), titanium nitride-covered SS, hand-polished SS, and microdiamond-covered hardmetal) under identical conditions. Gold-covered as
Xiaohua Liu et al.
Journal of the American Society for Mass Spectrometry, 22(12), 2125-2136 (2011-10-15)
A new model has been developed to account for adduct formation on multiply charged peptides observed in negative ion electrospray mass spectrometry. To obtain a stable adduct, the model necessitates an approximate matching of apparent gas-phase basicity (GB(app)) of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service