Skip to Content
Merck
All Photos(1)

Key Documents

EMS0005

Sigma-Aldrich

SOLu-Trypsin Dimethylated

Dimethlyated, Proteomics Grade, recombinant, expressed in Pichia pastoris, ready-to-use solution

Synonym(s):

Dimethylated Recombinant Trypsin for Protein Digestion in Mass Spectrometry, Dimethylated rTrypsin, Mass Spectrometry Trypsin Dimethlyated, No chemotryptic activity

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€148.00
4 X 100 μG
€400.00

€148.00


Estimated to ship on21 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€148.00
4 X 100 μG
€400.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.78

€148.00


Estimated to ship on21 April 2025


Request a Bulk Order

recombinant

expressed in Pichia pastoris

Quality Level

grade

Proteomics Grade

form

ready-to-use solution

potency

>10,000 units/mg

specific activity

≥10,000 units/mg protein (Enzymatic activity)

suitability

suitable for mass spectrometry

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

General description

SOLu-Trypsin Dimethylated (EMS0005) is our exclusive, Advanced Proteomics Grade enzyme that is solution stable for mass spectrometry. Designed to be stable in solution when refrigerated, SOLu-Trypsin Dimethylated can be used immediately without preparation. Other forms of trypsin require thawing or reconstitution, and must be discarded if not used immediately. SOLu-Trypsin Dimethylated allows excess product to be saved for future use, thus eliminating unnecessary waste and cost. SOLu-Trypsin Dimethylated is formulated with a high-purity recombinant trypsin, free of chymotryptic activity, to ensure high fidelity digestion.

Features and Benefits

  • Ready to use - no preparation, such as reconstitution or thawing, is required
  • Fits seamlessly into established workflow - no need to modify protocols
  • Eliminates waste - remains stable in the refrigerator after use so there is no need to discard excess product
  • Recombinant, porcine sequence - no chymotryptic activity
  • Stable for short-term use at room temperature in an autosampler or on a liquid handling robot

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Evaluation of Recombinant, Chemically Treated Trypsin in Proteomics and Protein Characterization Assays

Questions

  1. What's the feature/benefit of EMS0005 over EMS0004? Thanks!

    1 answer
    1. The difference between SOLu-Trypsin Dimethylated (EMS0005) and SOLu-Trypsin (EMS0004) products is primarily the stability. The dimethylated form, EMS0005, is more resistant to autolysis over time.

      Please see the links below to review the datasheet for each product:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/296/535/ems0005bul.pdf
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/334/377/ems0004bul.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service