Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
SE.11
UNSPSC Code:
41116104
Pricing and availability is not currently available.
product line
Spectroquant®
Quality Segment
packaging
pkg of 1 unit
measuring range
320-1100 nm
technique(s)
Vis spectroscopy: suitable, photometry: suitable
compatibility
for use with Spectroquant® (Reagents), for use with Spectroquant® (Test kits)
detection method
photometric (Vis spectroscopy)
storage temp.
10-30°C
General description
The Spectroquant Prove 100 plus is the best choice for those who primarily use our broad range of Spectroquant Test Kits, or only perform Vis measurements. The Spectroquant Prove 100 plus provides high quality and great value for money for your daily analyses. The Spectroquant Prove plus family is our series of cutting-edge spectrophotometers for easy analysis of water, food and beverage, and environmental samples. All Spectroquant Prove plus instruments are pre-programmed for over 200 Test Kits and automatic bar code recognition ensures the correct method parameters are applied and all essential information such as lot number, expiry date, and automatic calibration updates are recorded, making the use of analytical Test Kits as easy as never before. VIS Spectrophotometer 4 nm spectral bandwidth Spectroquant®. The Spectroquant® Prove plus photometers are upgraded highlighting smoother functionality with instant responsiveness, a more receptive display with faster processing, and increased storage capacity.
Application
Spectroquant® Prove plus photometers finds application in
- Wastewater testing
- Drinking water testing
- Process water testing
- Environmental testing (groundwater, soil analysis)
- Food & Beverage testing (brewery methods, sugar colors, oil determinations, bottled water, etc.)
Features and Benefits
Pre-programmed to work seamlessly with more than 150 Spectroquant® test kits, Spectroquant® Prove plus photometers are among the most trusted for your analysis
- Enhanced functionality for seamless operation and instant responsiveness
- Improved display responsiveness and faster processing capabilities
- Expanded storage capacity, allowing for storage of up to 7000 standard measurement results
- Extended warranty period of 2 years
- Easy to use and pre-programmed with pre-defined reagents
- Reduced use of chemicals compared to traditional methods
- Suitable for a broad measuring range per parameter (minimizes dilution work)
- Pre-programmed for over 300 analysis methods
- Instrument-supported AQA system for easy quality assurance
- Automatic cell size recognition for cell & reagent tests
- Live ID barcode for all cell & reagent tests (incl. lot and expiry data)
- No adapter is needed for the measurement of different cell sizes
- Kits can be conveniently stored at room temperature
Legal Information
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.