Skip to Content
Merck
All Photos(1)

Key Documents

762458

Sigma-Aldrich

(R)-(DTBM-SEGPHOS®)CuH solution

0.001 M in toluene

Synonym(s):

Hydro[(4R)-[4,4′-bi-1,3-benzodioxole]-5,5′-diylbis [bis[3,5-bis(1,1-dimethylethyl)-4-methoxyphenyl]phosphine-P]]copper(I) solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H101CuO8P2
CAS Number:
Molecular Weight:
1244.08
EC Number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

liquid

reaction suitability

core: copper
reagent type: catalyst

concentration

0.001 M in toluene

density

0.879 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Cu].COc1c(cc(cc1C(C)(C)C)P(c2cc(c(OC)c(c2)C(C)(C)C)C(C)(C)C)c3ccc4OCOc4c3-c5c6OCOc6ccc5P(c7cc(c(OC)c(c7)C(C)(C)C)C(C)(C)C)c8cc(c(OC)c(c8)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/C74H100O8P2.Cu/c1-67(2,3)47-33-43(34-48(61(47)75-25)68(4,5)6)83(44-35-49(69(7,8)9)62(76-26)50(36-44)70(10,11)12)57-31-29-55-65(81-41-79-55)59(57)60-58(32-30-56-66(60)82-42-80-56)84(45-37-51(71(13,14)15)63(77-27)52(38-45)72(16,17)18)46-39-53(73(19,20)21)64(78-28)54(40-46)74(22,23)24;/h29-40H,41-42H2,1-28H3;

InChI key

VXPZLAJMRQYNLT-UHFFFAOYSA-N

General description

Learn More at the Professor and Product Portal of Professor Bruce Lipshutz.

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136
SEGPHOS is a registered trademark of Takasago Intl. Corp.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F

Flash Point(C)

6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Designer surfactants enable diverse transformations like Suzuki-Miyaura, Olefin Metathesis, and 1,4-Addition to Enones in water by Prof. Bruce Lipshutz.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service