Skip to Content
Merck
All Photos(1)

Documents

449539

Sigma-Aldrich

Chlorocyclopentadienyl[(4S,5S)-2,2-dimethyl-α,α,α′,α′-tetraphenyl-1,3-dioxolane-4,5-dimethanolato]titanium

97%

Synonym(s):

(4S,5S)-Chloro-cyclopentadienyl-[2,2-dimethyl-1,3-dioxolan-4,5-bis(diphenylmethoxy)]titanium, (S,S)-Duthaler-Hafner reagent, [(4S,5S)-2,2-Dimethyl-1,3-dioxolan-4,5-bis(diphenylmethoxy)]cyclopentadienyl-chlorotitanium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H33ClO4Ti
CAS Number:
Molecular Weight:
612.96
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D +246°, c = 1 in chloroform

mp

209-213 °C (lit.)

SMILES string

Cl[Ti].[CH]1[CH][CH][CH][CH]1.CC2(C)O[C@@H]([C@H](O2)C(O)(c3ccccc3)c4ccccc4)C(O)(c5ccccc5)c6ccccc6

Application

A novel highly enantioselective allyl-transfer reagent. Excellent regio-, diastereo-, and enantioselectivities (usually >95% ee, >95% de) are obtained for the addition of allyl nucleophiles to aldehydes. Although this reagent is used in stoichiometric amounts, the chiral ligand and monocyclopentadienyltitanium trichloride can be easily recovered.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duthaler, R.O. Hafner, A.
Chemical Reviews, 92, 807-807 (1992)
Duthaler, R.O. et al.
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 64, 1897-1897 (1990)
Duthaler, R.O. et al.
Inorgorganica Chimica Acta, 222, 95-95 (1994)
Duthaler, R.O. et al.
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 62, 631-631 (1990)
A. Hafner et al.
Journal of the American Chemical Society, 114, 2321-2321 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service