Merck
  • Density dependence of retention factors of trans-stilbene oxide for chiral separation by supercritical fluid chromatography.
Density dependence of retention factors of trans-stilbene oxide for chiral separation by supercritical fluid chromatography.
Journal of chromatography. A (2017-11-05)
Toshitaka Funazukuri, Yuna Ono, Junichi Sakabe, Chang Yi Kong
ZUSAMMENFASSUNG
Retention factors for two enantiomers of trans-stilbene oxide, k
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3,5-Tri-tert-butylbenzol, 97%