Merck
Influence of Mn valence state and characteristic of TiO
Environmental technology (2017-01-04)
Dong Wook Kwon, Geo Jong Kim, Jong Min Won, Sung Chang Hong
ZUSAMMENFASSUNG
The effects of physicochemical properties of Mn-Ti catalysts on O