Merck
  • Didox and resveratrol sensitize colorectal cancer cells to doxorubicin via activating apoptosis and ameliorating P-glycoprotein activity.
Didox and resveratrol sensitize colorectal cancer cells to doxorubicin via activating apoptosis and ameliorating P-glycoprotein activity.
Scientific reports (2016-11-15)
Sahar A Khaleel, Ahmed M Al-Abd, Azza A Ali, Ashraf B Abdel-Naim
ZUSAMMENFASSUNG
Doxorubicin (DOX) has limited efficacy in colorectal cancer due to multi-drug resistance. Resveratrol (RES) and didox (DID) are polyhydroxyphenols with potential chemosensitizing effects. Herein, we assessed the chemomodulatory effects of RES and DID to DOX in colorectal cancer cells. Equitoxic combination of DOX with RES and DID in HCT 116 reduced the IC
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Didox, ≥98% (HPLC)