Merck
Construction of ICG encapsulated W
Materials science & engineering. C, Materials for biological applications (2017-09-04)
Hui-Quan Wang, Peng-Fei Hu, Yu Zheng, Zhe Zhao, Bin Zheng, Jin Chang, Han-Jie Wang, Jin-Hai Wang
ZUSAMMENFASSUNG
Photothermal therapy (PTT) has drawn tremendous attention because of its high therapeutic efficiency in targeting cells while minimizing the damage to normal tissues and organs. Tungsten oxide (W
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wolfram(IV)-chlorid, 95%