Merck
  • TLR4 signaling induces the release of microparticles by tumor cells that regulate inflammatory cytokine IL-6 of macrophages via microRNA let-7b.
TLR4 signaling induces the release of microparticles by tumor cells that regulate inflammatory cytokine IL-6 of macrophages via microRNA let-7b.
Oncoimmunology (2012-08-31)
Dapeng Li, Haibo Jia, Huafeng Zhang, Meng Lv, Jing Liu, Yi Zhang, Tao Huang, Bo Huang
ZUSAMMENFASSUNG
Tumor cells expressing TLRs is generally recognized to mediate tumor inflammation. However, whether and how tumor TLR signaling pathways negatively regulate tumor inflammation remains unclear. In this report, we find that TLR4 signaling of H22 hepatocarcinoma tumor cells is transduced through MyD88 pathway to actin cytoskeletons, leading to the release of microparticles (MPs), the cellular membrane-derived vesicles. As a result, tumor macrophages take up MPs and acquire MP-contained microRNA let-7b, which attenuates tumor inflammation by targeting proinflammatory cytokine IL-6. Thus, tumor TLR signaling, contrary to the original promoting effect, may play an opposite role in downregulating tumor inflammation through MP pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Latex beads, polystyrene, 3.0 μm mean particle size