Merck
  • Synthesis and biological evaluation of a novel phenyl substituted sydnone series as potential antitumor agents.
Synthesis and biological evaluation of a novel phenyl substituted sydnone series as potential antitumor agents.
Bioorganic & medicinal chemistry letters (2003-11-13)
Christopher S Dunkley, Charles J Thoman
ZUSAMMENFASSUNG
A series of compounds containing an N-(4'-substituted-3'-nitrophenyl)sydnone moiety with potential antitumor activity was prepared based on active analogues. The rationale behind the design of these compounds is presented along with the 4-step synthetic route to the derivatives in the 4'-position of the phenyl sydnone framework. Out of the six novel compounds, the 4'-fluoro derivative has an improved activity against all three cell lines as compared to the earlier leads.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trimethylenimin -hydrochlorid, 97%