- Regulation of TRPML1 function.
Regulation of TRPML1 function.
Biochemical Society transactions (2015-05-27)
Helen Waller-Evans, Emyr Lloyd-Evans
PMID26009188
ZUSAMMENFASSUNG
TRPML1 is a ubiquitously expressed cation channel found on lysosomes and late endosomes. Mutations in TRPML1 cause mucolipidosis type IV and it has been implicated in Alzheimer's disease and HIV. However, the mechanisms by which TRPML1 activity is regulated are not well understood. This review summarizes the current understanding of TRPML1 activation and regulation.