Merck
  • A novel gene codes for a putative G protein-coupled receptor with an abundant expression in brain.
A novel gene codes for a putative G protein-coupled receptor with an abundant expression in brain.
FEBS letters (1996-10-07)
B F O'Dowd, T Nguyen, K R Lynch, L F Kolakowski, M Thompson, R Cheng, A Marchese, G Ng, H H Heng, S R George
ZUSAMMENFASSUNG
Following the cloning of the dopamine receptors we continued a search of the human genome for related genes. We searched an EST data base and discovered cDNA fragments encoding novel G protein-coupled receptor genes. The available GenBank sequence of one of these EST fragments showed that it encoded a receptor with closest similarity to the D2 dopamine and adrenergic receptors. This cDNA was used to isolate the gene (GPR19), and the encoded receptor also demonstrated similarity with the neuropeptide Y receptor. The gene was mapped to chromosome 12, in region p13.2-12.3. Northern blot analysis revealed expression of GPR19 in peripheral regions, and brain regions significantly overlapping with the D2 receptor gene expression. A sequence of the rat orthologue of GPR19 was obtained and in situ hybridization analysis demonstrated a very abundant expression in rat brain.