Merck
  • Overexpression of cytosolic malate dehydrogenase (MDH2) causes overproduction of specific organic acids in Saccharomyces cerevisiae.
Overexpression of cytosolic malate dehydrogenase (MDH2) causes overproduction of specific organic acids in Saccharomyces cerevisiae.
Applied microbiology and biotechnology (1997-08-01)
O Pines, S Shemesh, E Battat, I Goldberg
ZUSAMMENFASSUNG
Saccharomyces cerevisiae accumulates L-malic acid through a cytosolic pathway starting from pyruvic acid and involving the enzymes pyruvate carboxylase and malate dehydrogenase. In the present study, the role of malate dehydrogenase in the cytosolic pathway was studied. Overexpression of cytosolic malate dehydrogenase (MDH2) under either the strong inducible GAL10 or the constitutive PGK promoter causes a 6- to 16-fold increase in cytosolic MDH activity in growth and production media and up to 3.7-fold increase in L-malic acid accumulation in the production medium. The high apparent Km of MDH2 for L-malic acid (11.8 mM) indicates a low affinity of the enzyme for this acid, which is consistent with the cytosolic function in the enzyme and differs from the previously published Km of the mitochondrial enzyme (MDH1, 0.28 mM). Under conditions of MDH2 overexpression, pyruvate carboxylase appears to be a limiting factor, thus providing a system for further metabolic engineering of L-malic acid production. The overexpression of MDH2 activity also causes an evaluation in the accumulation of fumaric acid and citric acid. Accumulation of fumaric acid is presumably caused by high intracellular L-malic acid concentrations and the activity of the cytosolic fumarase. The accumulation of citric acid may suggest the intriguing possibility that cytosolic L-malic acid is a direct precursor of citric acid in yeast.