Merck
L-cysteine-derived ambidextrous gelators of aromatic solvents and ethanol/water mixtures.
Langmuir : the ACS journal of surfaces and colloids (2013-01-25)
Amrita Pal, Joykrishna Dey
ZUSAMMENFASSUNG
A series of L-cysteine-derived double hydrocarbon chain amphiphilic gelators L-(3-alkyl-carbamoylsulfanyl)-2-(3-alkylurido)propionic acid with different hydrocarbon chain lengths (C6-C16) was designed and synthesized. These gelators efficiently gelate only aromatic solvents. The gelation ability increased with the increase of chain length up to C14, but then it dropped with further increase of chain length. The C12 and C14 derivatives also gelled ethanol/water mixtures. The gels were characterized by a number of methods, including FT-IR, NMR, and XRD spectroscopy, electron microscopy, and rheology. The amphiphiles were observed to form either flat lamellar or ribbonlike aggregates in aromatic solvents as well as in ethanol/water mixtures. The gelation in all the solvents employed was observed to be thermoreversible. The gel-to-sol transition temperature as well as mechanical strength of the organogels were observed to increase with the hydrocarbon chain length. Both types of gels of C8-C16 amphiphiles have gel-to-sol transition temperatures above the physiological temperature (310 K). FT-IR and variable temperature (1)H NMR measurements suggested that van der Waals interactions have major contribution in the gelation process. The gel-to-sol transition temperature and mechanical strength of the organogels in ethanol/water mixtures was observed to be higher than those of benzene organogel.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hexadecylisocyanat, 97%