Merck
TWEAK induces angiogenesis and proliferation of endothelial cells.
The Journal of biological chemistry (1999-03-20)
C N Lynch, Y C Wang, J K Lund, Y W Chen, J A Leal, S R Wiley
ZUSAMMENFASSUNG
TWEAK is a recently described member of the Tumor Necrosis Factor (TNF) ligand family whose transcripts are present in a wide variety of human tissues (Chicheportiche, Y., Bourdon, P. R., Xu, H., Hsu Y. M., Scott, H., Hession, C., Garcia, I., and Browning, J. L. (1997) J. Biol. Chem. 272, 32401-32410). TWEAK is a weak inducer of apoptosis in transformed cells when administered with interferon-gamma or cycloheximide (Chicheportiche, Y., Bourdon, P. R., Xu, H., Hsu Y. M., Scott, H., Hession, C., Garcia, I., and Browning, J. L. (1997) J. Biol. Chem. 272, 32401-32410; Masters, S. A., Sheridan, J. P., Pitti, R. M., Brush, A. G., and Ashkenazi, A. (1998) Curr. Biol. 8, 525-528) and also promotes IL-8 secretion in cultured cells. We report here that picomolar concentrations of recombinant soluble TWEAK induce proliferation in a variety of normal human endothelial cells and in aortic smooth muscle cells and reduce culture requirements for serum and growth factors. Blocking antibodies to Vascular Endothelial Growth Factor (VEGF) do not significantly inhibit TWEAK-induced proliferation, indicating that TWEAK does not function indirectly through up-regulation of VEGF. Pellets containing TWEAK induce a strong angiogenic response when implanted in rat corneas, suggesting a role for TWEAK in vasculature formation in vivo.