Merck
  • Synthesis, photo-physical and DFT studies of ESIPT inspired novel 2-(2',4'-dihydroxyphenyl) benzimidazole, benzoxazole and benzothiazole.
Synthesis, photo-physical and DFT studies of ESIPT inspired novel 2-(2',4'-dihydroxyphenyl) benzimidazole, benzoxazole and benzothiazole.
Journal of fluorescence (2013-05-18)
Vikas S Patil, Vikas S Padalkar, Abhinav B Tathe, Vinod D Gupta, N Sekar
ZUSAMMENFASSUNG
Novel ESIPT inspired benzimidazole, benzoxazole and benzothiazole were synthesized from 2,4-dihydroxy benzoic acid and 1,2-phenelenediamine, 2-aminophenol, and 2-aminothiophenol respectively. The synthesized 2-(2',4'-dihydroxyphenyl) benzimidazole, benzoxazole and benzothiazole are fluorescent and the emission characteristic are very sensitive to the micro-environment. They show a single absorption and dual emission with large Stokes shift originating from excited state intramolecular proton transfer. The absorption-emission characteristics of all these compounds are studied as a function of pH. The change in the electronic transition, energy levels, and orbital diagrams of synthesized compounds were investigated by the molecular orbital calculation and were correlated with the experimental spectral emission. Experimental absorption and emission wavelengths are in good agreement with those predicted using the Density Functional Theory (DFT) and Time-Dependent Density Functional Theory (TD-DFT) [B3LYP/6-31G(d)].
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Aminothiophenol, 99%