Merck
6-(Alkylamino)-9-benzyl-9H-purines. A new class of anticonvulsant agents.
Journal of medicinal chemistry (1988-03-01)
J L Kelley, M P Krochmal, J A Linn, E W McLean, F E Soroko
ZUSAMMENFASSUNG
Several 9-alkyl-6-substituted-purines were synthesized and tested for anticonvulsant activity against maximal electroshock-induced seizures (MES) in rats. Most compounds were prepared in three steps from 5-amino-4,6-dichloropyrimidine or in two steps via alkylation of 6-chloropurine. Potent anticonvulsant activity against MES resided in compounds that contain a benzyl substituent at the 9-position of 6-(methylamino)- or 6-(dimethyl-amino)purine. Among commonly used agents for control of seizures, this type of structure represents a new class of potent anticonvulsant agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Amino-4,6-dichlorpyrimidin, 97%