Merck
Concise total syntheses of variolin B and deoxyvariolin B.
The Journal of organic chemistry (2005-07-30)
Regan J Anderson, Jonathan B Hill, Jonathan C Morris
ZUSAMMENFASSUNG
The total synthesis of the marine alkaloid variolin B has been achieved in 8 steps and 17% overall yield, starting from commercially available 4-chloro-2-methylthiopyrimidine. The key reaction involves the tandem deoxygenation and cyclization of a triarylmethanol using a combination of triethylsilane and trifluoroacetic acid. In addition, the deoxygenated analogue was prepared in 6 steps and 23% overall yield, starting from the same starting material.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Chlor-2-methylthiopyrimidin, 98%