Merck
  • Cloning of a beta1 tubulin cDNA from an insect endocrine gland: developmental and hormone-induced changes in mRNA expression.
Cloning of a beta1 tubulin cDNA from an insect endocrine gland: developmental and hormone-induced changes in mRNA expression.
Molecular and cellular endocrinology (1998-09-02)
R Rybczynski, L I Gilbert
ZUSAMMENFASSUNG
A rapid increase in ecdysteroid hormone synthesis results when the insect prothoracic gland is stimulated with prothoracicotropic hormone (PTTH), a brain neuropeptide hormone. PTTH also stimulates the specific synthesis of several proteins, one of which is a beta tubulin. To further understand the possible roles of beta tubulin in the prothoracic gland, beta tubulin cDNA clones were isolated from a tobacco hornworm (Manduca sexta) gland cDNA library. Sequence analysis indicated that these clones were assignable to the beta1 tubulin isoform. Gland beta1 tubulin mRNA levels during the last larval instar and early pupal-adult development exhibited peaks that coincided with peaks in ecdysteroid synthesis. Manipulations of the glands hormonal milieu showed that beta1 tubulin mRNA levels respond to 20 hydroxyecdysone and PTTH. The data also support our earlier proposal that the prothoracic gland beta1 tubulin gene is ubiquitously expressed but exhibits tissue- and developmental-specific regulation of transcription and translation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
S-Hydropren, PESTANAL®, analytical standard