Merck
  • Contact dermatitis from dibutylthiourea. Report of a case with fine structural observations of epicutaneous testing with dibutylthiourea.
Contact dermatitis from dibutylthiourea. Report of a case with fine structural observations of epicutaneous testing with dibutylthiourea.
Contact dermatitis (1984-03-01)
L Kanerva, R Jolanki, M Plosila, T Estlander
ZUSAMMENFASSUNG
A 35-year-old machinist developed hand eczema from contact allergy to dibutylthiourea, a compound found in a paint and glue remover. Light and electron microscopy of his positive patch test was consistent with allergic contact dermatitis, showing Langerhans cell-mononuclear cell contacts in the epidermis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N′-Dibutylthioharnstoff, 97%