Merck
  • Manganese superoxide dismutase (SOD2)-mediated delayed radioprotection induced by the free thiol form of amifostine and tumor necrosis factor alpha.
Manganese superoxide dismutase (SOD2)-mediated delayed radioprotection induced by the free thiol form of amifostine and tumor necrosis factor alpha.
Radiation research (2007-03-29)
Jeffrey S Murley, Yasushi Kataoka, Kenneth L Baker, Alan M Diamond, William F Morgan, David J Grdina
ZUSAMMENFASSUNG
RKO36 cells, a subclone of RKO colorectal carcinoma cells that have been stably transfected with the pCMV-EGFP2Xho vector, were grown to confluence and then exposed to either the radioprotector WR-1065, i.e. the active thiol form of amifostine, for 30 min at doses of 40 microM and 4 mM or the cytokine tumor necrosis factor alpha (TNFalpha, TNFA) for 30 min at a concentration of 10 ng/ml and then washed. Total protein was isolated as a function of time up to 32 h after these treatments. Both doses of WR-1065 as well as the concentration of TNFalpha used were effective in elevating intracellular levels of the antioxidant protein SOD2 (also known as MnSOD) at least 15-fold over background levels as determined by Western blot analysis, while measured SOD2 activity was elevated between 5.5- and 6.9-fold. SOD2 reached a maximal level 24 h and 20 h after WR-1065 and TNFalpha treatments, respectively. The antioxidant proteins catalase and glutathione peroxidase (GPX) were also monitored over the 32-h period. In contrast to the robust changes observed in intracellular levels of SOD2 as a function of time after exposure of cells to WR-1065, catalase levels were elevated only 2.6-fold over background as determined by Western blot analysis, while GPX activity was unaffected by WR-1065 exposure. GPX protein levels were extremely low in cells, and analysis of GPX activity using a spectrophotometric method based on the consumption of reduced NADPH also revealed no measurable change as a function of WR-1065 or TNFalpha exposure. RKO36 cells either were irradiated with X rays in the presence of either 40 microM or 4 mM WR-1065 or 10 ng/ml TNFalpha or were irradiated 24 or 20 h later, respectively, when SOD2 protein levels were most elevated. The concentrations and exposure conditions used for WR-1065 and TNFalpha were not cytotoxic and had no effect on plating efficiencies or cell survival compared to untreated controls. No protection or sensitization was observed for cells irradiated in the presence of 40 microM WR-1065 or TNFalpha. Survival was elevated 1.90-fold for cells irradiated in the presence of 4 mM WR-1065. When RKO36 cells were irradiated with 2 Gy 24 h after 40 microM or 4 mM WR-1065 and 20 h after TNFalpha treatments when SOD2 levels were the most increased, survival was elevated 1.42-, 1.48- and 1.36-fold, respectively. This increased survival represents a SOD2-mediated delayed radioprotective effect. SOD2 appears to be an important antioxidant gene whose inducible expression is an important element in adaptive cellular responses in general, and the delayed radioprotective effect in particular. It can be induced by a range of agents including cytoprotective nonprotein thiols such as WR-1065 and pleiotropic cytokines such as TNFalpha.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
WR-1065, ≥98% (HPLC)