Merck
Conformational analysis of the anomeric forms of kojibiose, nigerose, and maltose using MM3.
Carbohydrate research (1992-06-16)
M K Dowd, J Zeng, A D French, P J Reilly
ZUSAMMENFASSUNG
Energy surfaces were computed for relative orientations of the relaxed pyranosyl rings of the two anomeric forms of kojibiose, nigerose, and maltose, the (1----2)-alpha, (1----3)-alpha, and (1----4)-alpha-linked D-glucosyl disaccharides, respectively. Twenty-four combinations of starting conformations of the rotatable side-groups were considered for each disaccharide. Optimized structures were calculated using MM3 on a 20 degree grid spacing of the torsional angles about the glycosidic bonds. The energy surfaces of the six disaccharides were similar in many respects but differed in detail within the low-energy regions. The maps also illustrate the importance of the exo-anomeric effect and linkage type in determining the conformational flexibility of disaccharides. Torsional conformations of known crystal structures of maltosyl-containing molecules lie in a lower MM3 energy range than previously reported.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kojibiose, ≥98% (HPLC)