Merck
  • Highly enantioselective synthesis of cyclopropylamine derivatives via Ru(II)-pheox-catalyzed direct asymmetric cyclopropanation of vinylcarbamates.
Highly enantioselective synthesis of cyclopropylamine derivatives via Ru(II)-pheox-catalyzed direct asymmetric cyclopropanation of vinylcarbamates.
Organic letters (2013-01-31)
Soda Chanthamath, Dao Thi Nguyen, Kazutaka Shibatomi, Seiji Iwasa
ZUSAMMENFASSUNG
The Ru(II)-Pheox-catalyzed asymmetric cyclopropanation of vinylcarbamates with diazoesters resulted in the corresponding cyclopropylamine derivatives in high yield and excellent diastereoselectivity (up to 96:4) and enantioselectivity (up to 99% ee).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cyclopropylamin, 98%