Merck
  • 25-Hydroxyvitamin D3 attenuates experimental periodontitis through downregulation of TLR4 and JAK1/STAT3 signaling in diabetic mice.
25-Hydroxyvitamin D3 attenuates experimental periodontitis through downregulation of TLR4 and JAK1/STAT3 signaling in diabetic mice.
The Journal of steroid biochemistry and molecular biology (2013-01-22)
Qi Wang, Hao Li, Honghui Xie, Min Fu, Bin Guo, Yi Ding, Wei Li, Haiyang Yu
ZUSAMMENFASSUNG
Vitamin D has been known to be closely associated with diabetes and periodontitis while the underlying mechanism has yet to be clarified. The present study aimed to discover the effect of 25-hydroxyvitamin D3 (25-OHD3) on glycemic control and periodontal health in mice with periodontitis superimposed on experimental diabetes (known as diabetic periodontitis). We showed that 25-OHD3 intraperitoneal injection attenuated diabetic periodontitis by reducing serum fasting blood glucose, glycosylated hemoglobin and TNF-α levels, which led to decreased alveolar bone loss. Immunohistochemical staining and western blot analysis of gingival epithelia revealed that vitamin D receptor (VDR) expression was enhanced upon 25-OHD3 treatment, while toll-like receptor 4 (TLR4) expression was reduced. The expressions of Janus family kinase (JAK) 1 and signal transducer and activator of transcription (STAT) 3 as well as their phosphorylation were inhibited in gingival epithelia of diabetic periodontitis mice, whereas the expression and phosphorylation of STAT1 remained unchanged. These results suggest that 25-OHD3 could improve diabetic periodontitis through downregulation of TLR4 and JAK1/STAT3 signaling in the gingival epithelium. Our study extends the previous findings on the regulation of diabetes with periodontitis, and may also provide a potential therapy for the patients with this disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-epi-25-Hydroxyvitamin D3, 98% (CP)