Merck
Transition-metal-free C-arylation at room temperature by arynes.
Organic letters (2012-07-27)
Ranjeet A Dhokale, Pramod R Thakare, Santosh B Mhaske
ZUSAMMENFASSUNG
A facile, fluoride-induced transition-metal-free chemoselective α-arylation of β-dicarbonyl compounds (malonamide esters) at room temperature using aryne intermediates has been demonstrated. Selective mono- or diarylation and generation of a quaternary benzylic stereocenter have also been achieved. The methodology will be highly useful for the synthesis of a library of CNS depressant barbiturate drugs like Phenobarbital.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Malonamid, 97%