- Chemical constituents from the radix of Curcuma wenyujin.
Chemical constituents from the radix of Curcuma wenyujin.
Fitoterapia (2009-05-19)
Zhong-Jun Ma, Zhao-Ke Meng, Peng Zhang
PMID19446613
ZUSAMMENFASSUNG
Phytochemical study on the ethanol extract of the radixes of Curcuma wenyujin Y. H. Chen et C. Ling led to the isolation of three new compounds curcuminol F (1) curcuminol G (2) and wenyujinoside (3) and a known compound aurantiamide (4). Their structures were elucidated on the basis of extensive spectroscopic analysis.