Merck
Practical synthesis of a p38 MAP kinase inhibitor.
The Journal of organic chemistry (2008-12-18)
Michał Achmatowicz, Oliver R Thiel, Philip Wheeler, Charles Bernard, Jinkun Huang, Robert D Larsen, Margaret M Faul
ZUSAMMENFASSUNG
p38 MAP kinase inhibitors have attracted considerable interest as potential agents for the treatment of inflammatory diseases. Herein, we describe a concise and efficient synthesis of inhibitor 1 that is based on a phthalazine scaffold. Highlights of our approach include a practical synthesis of a 1,6-disubstituted phthalazine building block 24 as well as the one-pot formation of boronic acid 27. Significant synthetic work to understand the reactivity principles of the intermediates helped in selection of the final synthetic route. Subsequent optimization of the individual steps of the final sequence led to a practical synthesis of 1.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phthalazin, 98%