Merck
Dicyclo-hex-yl[4-(dimethyl-amino)-phen-yl]phosphine selenide.
Acta crystallographica. Section E, Structure reports online (2012-01-20)
Zanele H Phasha, Sizwe Makhoba, Alfred Muller
ZUSAMMENFASSUNG
In the title mol-ecule, C(20)H(32)NPSe, the P atom has a distorted tetra-hedral environment resulting in an effective cone angle of 172°. Weak inter-molecular C-H⋯Se inter-actions are observed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dicyclohexyl(4-(N,N-dimethylamino)phenyl)phosphin, 95%