Merck
  • Synthesis, anti-inflammatory activity, and in vitro antitumor effect of a novel class of cyclooxygenase inhibitors: 4-(aryloyl)phenyl methyl sulfones.
Synthesis, anti-inflammatory activity, and in vitro antitumor effect of a novel class of cyclooxygenase inhibitors: 4-(aryloyl)phenyl methyl sulfones.
Journal of medicinal chemistry (2010-09-02)
Youssef Harrak, Giovanni Casula, Joan Basset, Glòria Rosell, Salvatore Plescia, Demetrio Raffa, Maria Grazia Cusimano, Ramon Pouplana, Maria Dolors Pujol
ZUSAMMENFASSUNG
Following our previous research on anti-inflammatory drugs (NSAIDs), we report on the design and synthesis of 4-(aryloyl)phenyl methyl sulfones. These substances were characterized for their capacity to inhibit cyclooxygenase (COX-1 and COX-2) isoenzymes. Molecular modeling studies showed that the methylsulfone group of these compounds was inserted deep in the pocket of the human COX-2 binding site, in an orientation that precludes hydrogen bonding with Arg120, Ser353, and Tyr355 through their oxygen atoms. The N-arylindole 33 was the most potent inhibitor of COX-2 and also the most selective (COX-1/COX-2 IC(50) ratio was 262). The indole derivative 33 was further tested in vivo for its anti-inflammatory activity in rats. This compound showed greater inhibitory activity than ibuprofen. Other compounds (20, 26, 9, and 30) showed strong activity against carrageenan-induced inflammation. The latter compounds showed a weak capacity to inhibit the proliferation of human cell lines K562, NCI-H460, and HT-29 in vitro.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%