Merck
Preparation and characterization of imprinted zeolite-Y for p-cresol removal in haemodialysis.
Materials science & engineering. C, Materials for biological applications (2019-07-28)
Yanuardi Raharjo, Ahmad Fauzi Ismail, Mohd Hafiz Dzarfan Othman, Nik Ahmad Nizam Nik Malek, Djoko Santoso
ZUSAMMENFASSUNG
In this work, the novel imprinted zeolite (IZ) was synthesized, and its properties and performance in terms of adsorption of p-Cresol, which represent the protein-bounded uremic toxins in aqueous phase under phosphate buffer saline, were studied and compared with the synthesized zeolite-Y (ZeoY-S) and commercial CBV 100 zeolite-Y (ZeoY-C). The ZeoY-S was synthesized from sodium aluminate, NaOH, H2O and SiO2 under aging for 24 h at room temperature and hydrothermal condition for 24 h at 100 °C, with an initial composition of 10SiO2:Al2O3:4Na2O:180H2O. The ZeoY-S has been modified by using the imprinting technology to produce the IZ via the use of p-Cresol as a template. The p-Cresol successfully imprinted on the zeolite-Y was proved through the multipoint Brunauer-Emmett-Teller (BET) and the performance of IZ that was compared to ZeoY-S and ZeoY-C. Based on the BET results, it proves that the pore size of IZ is in accordance with the target compound, which is p-Cresol at 0.79 nm. This modification was able to adsorb p-Cresol 2.5 and 3.5 times higher than ZeoY-S and ZeoY-C can, respectively. Langmuir and Freundlich adsorption isotherm models, together with the pseudo-first and -second order and intra-particle diffusion kinetics models, were used to investigate the adsorption behavior of p-Cresol on the zeolites. The IZ has 4.30 times greater competitive molecules than ZeoY-S and the properties of IZ were not influenced by the content of other phenolic group uremic toxins as competitive molecules. It can be concluded that the micropores of zeolite as adsorbent can be modified using the imprinting technology in order to increase its sensitivity and selectivity towards p-Cresol.