Merck
  • Effect of the interval from follicle aspiration to initiation of lengthened FSH treatment on follicular superstimulatory and superovulatory responses and embryo production in lactating Simmental cows.
Effect of the interval from follicle aspiration to initiation of lengthened FSH treatment on follicular superstimulatory and superovulatory responses and embryo production in lactating Simmental cows.
Theriogenology (2019-02-21)
Ümüt Cirit, Mehmet Ferit Özmen, İbrahim Küçükaslan, Mehmet Köse, Hüseyin Gökhan Kutsal, Elif Merve Çınar
ZUSAMMENFASSUNG
The present study evaluated follicular superstimulatory (FSS) and superovulatory (SOV) responses and in vivo embryo production in lactating Simmental cows treated with FSH starting 1 or 2 days after follicle aspiration (FA). The performance of a lengthened superovulation program, named 6dFSH-P36-hCG60, is described. At random stages of the estrous cycle, cows (n = 52) were subjected to ultrasound-guided transvaginal aspiration of all follicles ≥ 5 mm. After FA, cows were randomly assigned to one of two groups in which FSH treatments started 1 or 2 days after FA (groups FA-1D and FA-2D, respectively). Cows were superstimulated with a total of 500 μg pFSH over 6 days on a decreasing dose schedule and were pre-treated with a single dose of 400 IU of eCG 24 h before the start of FSH treatments. Follicular superstimulatory (the mean numbers of follicles ≥ 8 mm on the day of hCG treatment) and SOV responses (the mean numbers of CL and cows with ≥ 3 CL at the time of collection) were similar in FA-1D and FA-2D groups. However, when compared to FA-1D group, the number of unfertilized ova tended to decrease (0.4 vs 1.7; P = 0.065) and percentage of fertilized ova tended to increase (95.8% vs 84.6%; P = 0.066) in FA-2D group. Moreover, the mean numbers and percentages of both transferable embryos (8.0 and 77.6% vs 6.4 and 57.7%) and freezable embryos (5.3 and 51.5% vs 3.5 and 31.1%) were numerically higher in FA-2D group than FA-1D group. The results of the study suggest that starting a lengthened superovulation programs in Simmental cows 2 days after FA has potential to increase percentage of fertilized ova and the number of transferable and freezable embryos, although new studies may be needed to confirm this findings.