Merck
  • Neural progenitor cells derived from adult bone marrow mesenchymal stem cells promote neuronal regeneration.
Neural progenitor cells derived from adult bone marrow mesenchymal stem cells promote neuronal regeneration.
Life sciences (2012-09-25)
Yue Tang, Yong-Chun Cui, Xiao-Juan Wang, Ai-Li Wu, Guang-Fu Hu, Fu-Liang Luo, Jia-Kang Sun, Jing Sun, Li-Ke Wu
ZUSAMMENFASSUNG
It is well known that neural stem/progenitor cells (NS/PC) are an ideal cell type for the treatment of central nervous system (CNS) disease. However, ethical problems have severely hampered fetal NS/PC from being widely used as a source for stem cell therapy. Recently, it has been demonstrated that autologous bone marrow mesenchymal stem cells (BMSC) can transdifferentiate into neural progenitor cells (NPC). The biological function of BMSC derived NPC (MDNPC) in neuronal systems remains unknown. In the present study, we aimed to investigate whether MDNPC can promote in vitro neural regeneration, a process comprising mainly the generation of neurons and neurotransmitters. We co-cultured BMSC, MDNPC or fetal NS/PC with PC12 cells and studied their roles on proliferation, neurite formation and dopamine release from PC12 cells. Furthermore, we also explored the mechanisms by which MDNPC regulate dopamine secretion from PC12 derived neural cells using Western blot. We found that both MDNPC and NS/PC had similar morphologies and there were no significant differences between MDNPC and NS/PC in promoting PC12 cell proliferation, neurite outgrowth, and dopamine release. We also demonstrated that NS/PC induced dopamine secretion was associated with an upregulation of dopamine transporter (DAT) levels. In summary, MDNPC were comparable to NS/PC in promoting neural regeneration, indicating that MDNPC are a promising candidate source of neural stem cells for the treatment of neurological diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Neurite Outgrowth Assay Kit (1 µm), The NS225 Neurite Outgrowth Assay Kit (1 μm) is based on the use of Millicell cell culture inserts (chambers) containing a permeable membrane with 1 μm pores at the base.